Learn More
Medchemexpress LLC ZY-444 | 1802650-31-2 | 99.5% | C26H28N4OS | 100 MG
SDP
Catalog No. 5000354922
Encompass_Preferred
ZY-444 is an anti-cancer agent that targets pyruvate carboxylase (PC). It suppresses the Wnt/β-catenin/Snail signaling pathway by blocking the nuclear translocation of β-catenin. This compound selectively inhibits proliferation, migration, and invasion, and induces apoptosis in cancer cells. It has demonstrated potent anti-tumor effects in cancer mouse models and is suitable for the study of breast cancer, lung cancer (NSCLC), prostate cancer, and iodine-refractory thyroid cancer.
  • Suppresses Wnt/β-catenin/Snail signaling pathway
  • Inhibits proliferation, migration, and invasion in cancer cells
  • Induces apoptosis in cancer cells
  • Demonstrates anti-tumor effects in cancer mouse models
  • Can be used for breast, lung, prostate, and thyroid cancer research
Catalog No. 50-003-54922 Supplier Medchemexpress LLC Supplier No. HY142870100MG
ZY-444 is an anti-cancer agent that targets pyruvate carboxylase (PC). It suppresses the Wnt/β-catenin/Snail signaling pathway by blocking the nuclear translocation of β-catenin. This compound selectively inhibits proliferation, migration, and invasion, and induces apoptosis in cancer cells. It has demonstrated potent anti-tumor effects in cancer mouse models and is suitable for the study of breast cancer, lung cancer (NSCLC), prostate cancer, and iodine-refractory thyroid cancer.
  • Suppresses Wnt/β-catenin/Snail signaling pathway
  • Inhibits proliferation, migration, and invasion in cancer cells
  • Induces apoptosis in cancer cells
  • Demonstrates anti-tumor effects in cancer mouse models
  • Can be used for breast, lung, prostate, and thyroid cancer research