Learn More
Medchemexpress LLC ZY-444 | 1802650-31-2 | 99.5% | C26H28N4OS | 50 MG
SDP
Catalog No. 5000354921
Encompass_Preferred
ZY-444 is an anti-cancer agent that targets pyruvate carboxylase (PC). It suppresses the Wnt/β-catenin/Snail signaling pathway by blocking the nuclear translocation of β-catenin. This compound selectively inhibits the proliferation, migration, and invasion of cancer cells and induces apoptosis. It has demonstrated potent anti-tumor effects in cancer mouse models.
  • Selectively inhibits proliferation, migration, and invasion of cancer cells
  • Induces apoptosis in cancer cells
  • Can be utilized for research into breast cancer, lung cancer (NSCLC), prostate cancer, and iodine-refractory thyroid cancer
  • Demonstrates potent anti-tumor effects in cancer mouse models
Catalog No. 50-003-54921 Supplier Medchemexpress LLC Supplier No. HY14287050MG
ZY-444 is an anti-cancer agent that targets pyruvate carboxylase (PC). It suppresses the Wnt/β-catenin/Snail signaling pathway by blocking the nuclear translocation of β-catenin. This compound selectively inhibits the proliferation, migration, and invasion of cancer cells and induces apoptosis. It has demonstrated potent anti-tumor effects in cancer mouse models.
  • Selectively inhibits proliferation, migration, and invasion of cancer cells
  • Induces apoptosis in cancer cells
  • Can be utilized for research into breast cancer, lung cancer (NSCLC), prostate cancer, and iodine-refractory thyroid cancer
  • Demonstrates potent anti-tumor effects in cancer mouse models