Learn More
TARGETMOL CHEMICALS INC ZY-444 5MG
SDP
Catalog No. 502880112
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ZY-444 is a small molecule inhibiting the progression of breast cancer by targeting pyruvate carboxylase. Purity 99.14%
Catalog No. 50-288-0112 Supplier TARGETMOL CHEMICALS INC Supplier No. T626205MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ZY-444 is a small molecule inhibiting the progression of breast cancer by targeting pyruvate carboxylase. Purity 99.14%