Learn More
Med Vet International Zydaclin Tablets (Clindamycin Hydrochloride), 25mg, 400 Count
Catalog No. 502828151
Encompass_Preferred
ZydaClin Tablets (clindamycin hydrochloride) are indicated for the treatment of infections caused by susceptible strains of the designated microorganisms in the specific conditions listed below: Dogs: Skin infections (wounds and abscesses) due to coagulase positive staphylococci (Staphylococcus aureus or Staphylococcus intermedius). Deep wounds and abscesses due to Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens. Dental infections due to Staphyloccus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens. Osteomyelitis due to Staphylococcus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens.
Catalog No. 50-282-8151 Supplier Med Vet International Supplier No. RXZYDA25400V
ZydaClin Tablets (clindamycin hydrochloride) are indicated for the treatment of infections caused by susceptible strains of the designated microorganisms in the specific conditions listed below: Dogs: Skin infections (wounds and abscesses) due to coagulase positive staphylococci (Staphylococcus aureus or Staphylococcus intermedius). Deep wounds and abscesses due to Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens. Dental infections due to Staphyloccus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens. Osteomyelitis due to Staphylococcus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens.