Learn More
Med Vet International Zydaclin (Clindamycin) Oral Drops For Dogs And Cats, 20mL, Each
Catalog No. 502828152
Encompass_Preferred
ZydaClin Oral Drops liquid (for use in dogs and cats) is a palatable formulation intended for oral administration of clindamycin. Each mL of ZydaClin Oral Drops liquid contains clindamycin hydrochloride equivalent to 25 mg clindamycin; and ethyl alcohol, 8.64%. ZydaClin Oral Drops liquid is non-refrigerated and highly effective, with multiple indications for both dogs and cats. Effective in the treatment of various tissue infections, dental infections and osteomyelitis. It is generally well tolerated by the animal and presented in a ready-to-use vial with included dropper for accurate, safe administration.
Catalog No. 50-282-8152 Supplier Med Vet International Supplier No. RXZYDADROP2520V
ZydaClin Oral Drops liquid (for use in dogs and cats) is a palatable formulation intended for oral administration of clindamycin. Each mL of ZydaClin Oral Drops liquid contains clindamycin hydrochloride equivalent to 25 mg clindamycin; and ethyl alcohol, 8.64%. ZydaClin Oral Drops liquid is non-refrigerated and highly effective, with multiple indications for both dogs and cats. Effective in the treatment of various tissue infections, dental infections and osteomyelitis. It is generally well tolerated by the animal and presented in a ready-to-use vial with included dropper for accurate, safe administration.