Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL
Catalog No. 14222908
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Standard, Round
Filtered:
Filtered
Non-filtered
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Tip Style Filtered
14-222-908 Sterile Standard, Round Non-filtered
14-222-907 Non-sterile Standard, Round Non-filtered
14-222-922 Sterile Standard, Round Filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 14-222-908 Supplier Axygen™ Supplier No. ZT100RS
Add to Cart
Add to Cart
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Clear, autoclavable tips; color-coded racks.
  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Gray
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Standard, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polypropylene