Learn More
Zymo Research Corporation Zymoclean™ Gel RNA Recovery Kit (50 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. NC1027310
Encompass_Preferred
The procedure combines a unique, single-step agarose dissolving RAD Buffer with Zymo-Spin™ Column technology to yield high quality, purified RNA in just minutes. The purified RNA is eluted with DNase/RNase-Free Water into small volumes and is highly concentrated and suitable for subsequent RNA-based manipulations including RT-PCR.
Catalog No. NC1027310 Supplier Zymo Research Corporation Supplier No. R1011
The procedure combines a unique, single-step agarose dissolving RAD Buffer with Zymo-Spin™ Column technology to yield high quality, purified RNA in just minutes. The purified RNA is eluted with DNase/RNase-Free Water into small volumes and is highly concentrated and suitable for subsequent RNA-based manipulations including RT-PCR.