Learn More
Zymo Research Corporation DNA Clean & Concentrator™-5 (200 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. NC9552153
Encompass_Preferred
The DNA Clean & Concentrator-5 (DCC-5) can also be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield DNA that is free of salts and contaminants injust 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.
Catalog No. NC9552153 Supplier Zymo Research Corporation Supplier No. D4014
The DNA Clean & Concentrator-5 (DCC-5) can also be used for post-RT cDNA clean-up and purification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield DNA that is free of salts and contaminants injust 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.