Learn More
Zymo Research Corporation Quick-DNA™ Miniprep Plus Kit (50 Preps)
SDP
Catalog No. NC0933233
Encompass_Preferred
The Quick-DNA™ Miniprep Plus Kit facilitates rapid and efficient purification of DNA from any biological fluids, cultured/monolayer cells, or solid tissues by combining enzymatic and chemical extraction regimens. Innovative reagents and Zymo-Spin™ Technology allow for ultra-pure and concentrated genomic DNA 50 kb to be eluted in as little as 35 l. Zymo-Spin™ Columns ensure no buffer retention. Purified DNA is RNA-free, bypassing the need for RNase A treatment and ensuring accurate quantification forapplications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.
Catalog No. NC0933233 Supplier Zymo Research Corporation Supplier No. D4068
The Quick-DNA™ Miniprep Plus Kit facilitates rapid and efficient purification of DNA from any biological fluids, cultured/monolayer cells, or solid tissues by combining enzymatic and chemical extraction regimens. Innovative reagents and Zymo-Spin™ Technology allow for ultra-pure and concentrated genomic DNA 50 kb to be eluted in as little as 35 l. Zymo-Spin™ Columns ensure no buffer retention. Purified DNA is RNA-free, bypassing the need for RNase A treatment and ensuring accurate quantification forapplications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.