Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zymo Research Corporation DNA Digestion Buffer, 16 ml
SDP

Catalog No. NC1799127
Encompass_Preferred

DNA Digestion Buffer 16ml

Catalog No. NC1799127 Supplier Zymo Research Corporation Supplier No. E1010116
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA Digestion Buffer 16ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.