Learn More
Zymo Research Corporation IPTG (Isopropyl-β-D-thiogalactopyranoside), 5x5 ml
SDP
Catalog No. 501957642
Encompass
The IPTG solution can be used in conjunction with X-Gal for blue-white colony screening or by itself to induce expression of recombinant genes under control of the lac operon.
Catalog No. 50-195-7642 Supplier Zymo Research Corporation Supplier No. I100125
The IPTG solution can be used in conjunction with X-Gal for blue-white colony screening or by itself to induce expression of recombinant genes under control of the lac operon.