Learn More
Zymo Research Corporation M-Elution Buffer, 4 ml
SDP
Catalog No. NC1053964
Encompass_Preferred
M-Elution Buffer (4 ml). For use with EZ DNA Methylation™ bisulfite conversion Kits. Sold individually or as part of a bisulfite conversion kit.
Catalog No. NC1053964 Supplier Zymo Research Corporation Supplier No. D50026
M-Elution Buffer (4 ml). For use with EZ DNA Methylation™ bisulfite conversion Kits. Sold individually or as part of a bisulfite conversion kit.