Learn More
Research Products International Corp Zymo Quick-DNA/RNA MagBead Kit, 96 Preps
SDP
Catalog No. 501977217
Encompass
The Quick-DNA/RNA MagBead Kit provides a high-throughput, magnetic-bead based purification of both high-quality DNA and total RNA (including small RNAs) from the same starting sample. The provided DNA/RNA Shield inactivates infectious agent and is ideal for sample storage at ambient temperatures. The extraction method has the option to recover total nucleic acids in one elution or DNA & RNA in separate elutions without the use of phenol. It is eluted into ≥50 µl of DNase/RNase-free water and is ready for any downstream application including RT-PCR, sequencing, etc.
Quick & Easy: Extract DNA and RNA from any sample including cells, solid tissue, whole blood, biological liquids, FFPE tissue, environmental (plant/seed), swabs (stool, soil, microbial samples), etc.
Ultra-Pure: Ready for Next-generation sequencing, RT-qPCR, arrays, etc. DNase I included (Plus kit and MagBead only)
Catalog No. 50-197-7217 Supplier Research Products International Corp Supplier No. ZR2130
The Quick-DNA/RNA MagBead Kit provides a high-throughput, magnetic-bead based purification of both high-quality DNA and total RNA (including small RNAs) from the same starting sample. The provided DNA/RNA Shield inactivates infectious agent and is ideal for sample storage at ambient temperatures. The extraction method has the option to recover total nucleic acids in one elution or DNA & RNA in separate elutions without the use of phenol. It is eluted into ≥50 µl of DNase/RNase-free water and is ready for any downstream application including RT-PCR, sequencing, etc.
Quick & Easy: Extract DNA and RNA from any sample including cells, solid tissue, whole blood, biological liquids, FFPE tissue, environmental (plant/seed), swabs (stool, soil, microbial samples), etc.
Ultra-Pure: Ready for Next-generation sequencing, RT-qPCR, arrays, etc. DNase I included (Plus kit and MagBead only)