Learn More
Zymo Research Corporation Quick-DNA/RNA Viral MagBead (96 preps)
SDP
Catalog No. NC1799125
Encompass_Preferred
The kit also features a buffer system that facilitates complete viral particle lysis for efficient nucleic acid isolation. Small (> 50 nt) and large (> 200 kb) DNA and RNA are bound to magnetic beads, washed and eluted.The isolated high-quality nucleic acids are ready for all downstream applications such as Next-Gen sequencing, hybridization-based andRT/qPCR detection.
Catalog No. NC1799125 Supplier Zymo Research Corporation Supplier No. R2140
The kit also features a buffer system that facilitates complete viral particle lysis for efficient nucleic acid isolation. Small (> 50 nt) and large (> 200 kb) DNA and RNA are bound to magnetic beads, washed and eluted.The isolated high-quality nucleic acids are ready for all downstream applications such as Next-Gen sequencing, hybridization-based andRT/qPCR detection.