Learn More
Zymo Research Corporation Zymo-Seq RRBS Library Kit (24 preps)
SDP
Catalog No. 501957605
Encompass_Preferred
DNA methylation occurs predominantly in CpG contexts, and these CpG dinucleotides are more abundant in select regions of the genome. By enriching for CpG-dense regions and sequencing only the fragments pertaining to those regions, the RRBS platform allows for the capture of a significant amount of methylation data while reducing the amount of sequencing, leading to a substantially decreased cost. This combination makes RRBS the perfect platform for pilot studies. Libraries prepared by Zymo-Seq RRBS™ Library Kit cover 75% CpG islands, 70% gene promoters, 75% gene bodies, and 2.5-4 million unique CpG sites at 5-10x coverage when applied to human samples.
Catalog No. 50-195-7605 Supplier Zymo Research Corporation Supplier No. D5460
DNA methylation occurs predominantly in CpG contexts, and these CpG dinucleotides are more abundant in select regions of the genome. By enriching for CpG-dense regions and sequencing only the fragments pertaining to those regions, the RRBS platform allows for the capture of a significant amount of methylation data while reducing the amount of sequencing, leading to a substantially decreased cost. This combination makes RRBS the perfect platform for pilot studies. Libraries prepared by Zymo-Seq RRBS™ Library Kit cover 75% CpG islands, 70% gene promoters, 75% gene bodies, and 2.5-4 million unique CpG sites at 5-10x coverage when applied to human samples.