Learn More
Zymo Research Corporation Zymo-Seq™ UDI Primer Plate (Indexes 1-96)
SDP
Catalog No. 502037037
Encompass
Indexes come as pre-mixes, and the forward and reverse primers are provided at 5 M total concentration (2.5 M each). These indexes can be used to barcode any library containing TruSeq adapters and are compatible with the Zymo-Seq RiboFree™ Total RNA Library Kit (R3000 & R3003). Zymo-Seq™ index primers generate sequencing libraries that are compatible with all Illumina platforms.
Catalog No. 50-203-7037 Supplier Zymo Research Corporation Supplier No. D3096
Indexes come as pre-mixes, and the forward and reverse primers are provided at 5 M total concentration (2.5 M each). These indexes can be used to barcode any library containing TruSeq adapters and are compatible with the Zymo-Seq RiboFree™ Total RNA Library Kit (R3000 & R3003). Zymo-Seq™ index primers generate sequencing libraries that are compatible with all Illumina platforms.