Learn More
Zymo Research Corporation Zymo-Spin IC-XM Columns, 50 pack
SDP
Catalog No. NC1530030
Encompass_Preferred
The Zymo-Spin IC-XM features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 µg of DNA, and 10 µg of RNA, in ≥ 10 µl eluate. Capacity is 800 µl. Used in the Quick-DNA Microprep Plus kit (Cat. #D4074).
Catalog No. NC1530030 Supplier Zymo Research Corporation Supplier No. C110350
The Zymo-Spin IC-XM features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 µg of DNA, and 10 µg of RNA, in ≥ 10 µl eluate. Capacity is 800 µl. Used in the Quick-DNA Microprep Plus kit (Cat. #D4074).