Learn More
Zymo Research Corporation Zymo-Spin 384 Well Plate, 2 plate
SDP
Catalog No. 502037018
Encompass_Preferred
384-well polypropylene spin plate with 1.2 µm glass fiber membrane. 140 µl/well, long drip.
Catalog No. 50-203-7018 Supplier Zymo Research Corporation Supplier No. C2012
384-well polypropylene spin plate with 1.2 µm glass fiber membrane. 140 µl/well, long drip.