Learn More
Zymo Research Corporation Zymo-Spin™ I Columns (250 Pack) (Uncapped)
SDP
Catalog No. NC9235381
Encompass_Preferred
The Zymo-Spin I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g DNA or RNA in 6 l eluate. Capacity is 800 l.
Catalog No. NC9235381 Supplier Zymo Research Corporation Supplier No. C1003250
The Zymo-Spin I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g DNA or RNA in 6 l eluate. Capacity is 800 l.