Learn More
Zymo Research Corporation Zymo-Spin I-96-XL Plates, 2 plate
SDP
Catalog No. 502037016
Encompass_Preferred
Its deep-well, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 µg of DNA, or 10 µg of RNA, in ≥15 µl eluate per well. Capacity is 1.1 ml per well.
Catalog No. 50-203-7016 Supplier Zymo Research Corporation Supplier No. C2010
Its deep-well, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 µg of DNA, or 10 µg of RNA, in ≥15 µl eluate per well. Capacity is 1.1 ml per well.