Learn More
Zymo Research Corporation Zymo-Spin™ IB-96 Plate (Shallow-Well) (2 Plates)
SDP
Catalog No. 502037014
Encompass_Preferred
Its low-profile, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 g DNA or RNA in 15 l/well elution buffer or water. Opaque black in color. Capacity is 600 l per well.
Catalog No. 50-203-7014 Supplier Zymo Research Corporation Supplier No. C2006
Its low-profile, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 g DNA or RNA in 15 l/well elution buffer or water. Opaque black in color. Capacity is 600 l per well.