Learn More
Zymo Research Corporation Zymo-Spin™ IB Columns (250 Pack) (Uncapped)
SDP
Catalog No. 502036994
Encompass_Preferred
The Zymo-Spin IB features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g of DNA, and 10 g RNA, in 6 l eluate. Capacity is 800 l.
Catalog No. 50-203-6994 Supplier Zymo Research Corporation Supplier No. C1014250
The Zymo-Spin IB features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g of DNA, and 10 g RNA, in 6 l eluate. Capacity is 800 l.