Learn More
Zymo Research Corporation Zymo-Spin™ IC Columns (50 Pack) (Capped)
SDP
Catalog No. NC0749301
Encompass_Preferred
The Zymo-Spin IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g DNA or RNA in 6 l eluate. Capacity is 800 l.
Catalog No. NC0749301 Supplier Zymo Research Corporation Supplier No. C100450
The Zymo-Spin IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g DNA or RNA in 6 l eluate. Capacity is 800 l.