Learn More
Zymo Research Corporation Zymo-Spin™ IC Columns (250 Pack) (Capped)
SDP
Supplier:  Zymo Research Corporation C1004250
Encompass_Preferred
The Zymo-Spin IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g DNA or RNA in 6 l eluate. Capacity is 800 l.
Catalog No. NC9587302