Learn More
Research Products International Corp Zymo-Spin IC Columns, Capped, 250/pk
SDP
Supplier:  Research Products International Corp ZC1004250
Encompass
Capped version of the Zymo-Spin I column. The Zymo-Spin IC Fast-Spin column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 µg DNA or RNA in≥ 6 µl eluate. Capacity is 800 µl.
Catalog No. 50-197-7375