Learn More
Zymo Research Corporation Zymo-Spin™ IC-XL (50 Pack)
SDP
Catalog No. NC1301212
Encompass_Preferred
The Zymo-Spin IC-XL features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 10 g of DNA, and 20 g of RNA, in 10 l eluate. Capacity is 1 ml.
Catalog No. NC1301212 Supplier Zymo Research Corporation Supplier No. C100250
The Zymo-Spin IC-XL features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 10 g of DNA, and 20 g of RNA, in 10 l eluate. Capacity is 1 ml.