Learn More
Zymo Research Corporation Zymo Spin™ II-P (50 Pack)
SDP
Catalog No. 501251316
Encompass_Preferred
Features polypropylene construction and contains a unique silica-based matrix that allows purification of up to 100 g of plasmid DNA in 25 l eluate when used in combination with ZymoPURE™ Plasmid buffers. Capacity is 800 l.
Catalog No. 50-125-1316 Supplier Zymo Research Corporation Supplier No. C105550
Features polypropylene construction and contains a unique silica-based matrix that allows purification of up to 100 g of plasmid DNA in 25 l eluate when used in combination with ZymoPURE™ Plasmid buffers. Capacity is 800 l.