Learn More
Research Products International Corp Zymo-Spin II-PX Columns 50pk
SDP
Catalog No. 502388455
Encompass
Available as a refill for the ZymoPURE Plasmid Miniprep Kit. The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin II-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 100 µg of plasmid DNA in ≥ 25 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 800 µl.
Catalog No. 50-238-8455 Supplier Research Products International Corp Supplier No. ZC108650
Available as a refill for the ZymoPURE Plasmid Miniprep Kit. The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin II-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 100 µg of plasmid DNA in ≥ 25 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 800 µl.