Learn More
Zymo Research Corporation Zymo-Spin II-PX Columns (50 Pack)
SDP
Catalog No. NC2144347
Encompass_Preferred
Available as a refill for the ZymoPURE Plasmid Miniprep Kit. The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin II-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 100 µg of plasmid DNA in ≥ 25 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 800 µl.
This item is not returnable. View return policy
Catalog No. NC2144347 Supplier Zymo Research Corporation Supplier No. C108650
Available as a refill for the ZymoPURE Plasmid Miniprep Kit. The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin II-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 100 µg of plasmid DNA in ≥ 25 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 800 µl.