Learn More
Zymo Research Corporation Zymo-Spin™ IIC-XL (50 Pack)
SDP
Catalog No. 501957576
Encompass_Preferred
The Zymo-Spin IIC-XL features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 50 g of RNA, in 35 l eluate. Capacity is 900 l.
Catalog No. 50-195-7576 Supplier Zymo Research Corporation Supplier No. C110250
The Zymo-Spin IIC-XL features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 50 g of RNA, in 35 l eluate. Capacity is 900 l.