Learn More
Zymo Research Corporation Zymo-Spin IIC-XLR Columns, 50 pack
SDP
Catalog No. 501957578
Encompass_Preferred
The Zymo-Spin IIC-XLR features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 µg of DNA, and 50 µg of RNA, in ≥ 35 µl eluate. Capacity is 800 µl.
Catalog No. 50-195-7578 Supplier Zymo Research Corporation Supplier No. C110450
The Zymo-Spin IIC-XLR features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 µg of DNA, and 50 µg of RNA, in ≥ 35 µl eluate. Capacity is 800 µl.