Learn More
Zymo Research Corporation Zymo-Spin IICR Columns, 5x50 pack
SDP
Catalog No. 501957574
Encompass_Preferred
The Zymo-Spin IICR features durable polypropylene construction with an attached cap and contains a unique silica-based matrix that allows purification of up to 25 µg of DNA or 50 µg of RNA, in ≥ 25 µl eluate. Capacity is 900 µl.
Catalog No. 50-195-7574 Supplier Zymo Research Corporation Supplier No. C1078250
The Zymo-Spin IICR features durable polypropylene construction with an attached cap and contains a unique silica-based matrix that allows purification of up to 25 µg of DNA or 50 µg of RNA, in ≥ 25 µl eluate. Capacity is 900 µl.