Learn More
Zymo Research Corporation Zymo-Spin™ III Columns (250 Pack) (Uncapped)
SDP
Catalog No. 501251274
Encompass_Preferred
The Zymo-Spin III features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g DNA or RNA in 35 l eluate. Capacity is 800 l.
Catalog No. 50-125-1274 Supplier Zymo Research Corporation Supplier No. C1005250
The Zymo-Spin III features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g DNA or RNA in 35 l eluate. Capacity is 800 l.