Learn More
Zymo Research Corporation Zymo-Spin™ III Columns (50 Pack) (Uncapped)
SDP
Catalog No. 502036990
Encompass_Preferred
The Zymo-Spin III features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g DNA or RNA in 35 l eluate. Capacity is 800 l.
Catalog No. 50-203-6990 Supplier Zymo Research Corporation Supplier No. C100550
The Zymo-Spin III features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g DNA or RNA in 35 l eluate. Capacity is 800 l.