Learn More
Zymo Research Corporation Zymo-Spin III-HRC Filters (50 pack)
SDP
Supplier:  Zymo Research Corporation C105850
Encompass_Preferred
Designed for efficient removal of polyphenolic compounds, humic/fulvic acids, tannins, melanin, etc. from most impure DNA and RNA preparations. Must be prepared using Prep Solution before use.
Catalog No. 50-203-7010