Learn More
Zymo Research Corporation Zymo Spin™ III-P Column Assembly (5 Pack) w/ 15 ml and 50 ml Reservoir
SDP
Catalog No. 501251309
Encompass_Preferred
The Zymo-Spin III-P features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 300 g of plasmid DNA in 100 l eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.
Catalog No. 50-125-1309 Supplier Zymo Research Corporation Supplier No. C10405
The Zymo-Spin III-P features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 300 g of plasmid DNA in 100 l eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.