Learn More
Zymo Research Corporation Zymo Spin™ III-S Columns (25 Pack) w/ LuerLock Caps and 15 ml Reservoir
SDP
Catalog No. 501251313
Encompass_Preferred
Compatible on a microcentrifuge or vacuum manifold for the purification of cell-free DNA. Features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g of DNA in 35 l eluate. Capacity is 15 ml.
Catalog No. 50-125-1313 Supplier Zymo Research Corporation Supplier No. C104925
Compatible on a microcentrifuge or vacuum manifold for the purification of cell-free DNA. Features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 g of DNA in 35 l eluate. Capacity is 15 ml.