Learn More
Zymo Research Corporation Zymo-Spin™ IIICG Columns (250 Pack) (Capped) Green Retension Ring
SDP
Catalog No. 501251276
Encompass_Preferred
The Zymo-Spin IIICG features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 100 g of RNA, in 50l eluate. Capacity is 800 l.
Catalog No. 50-125-1276 Supplier Zymo Research Corporation Supplier No. C1006250G
The Zymo-Spin IIICG features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 100 g of RNA, in 50l eluate. Capacity is 800 l.