Learn More
Zymo Research Corporation Zymo-Spin™ IV Columns (50 Pack)
SDP
Catalog No. NC1094586
Encompass_Preferred
It is ideal for clarifying solutions, including crude cell lysates and homogenates. The silica filtration membrane has an approximate 10 - 20 m pore size. Capacity is 700 l.
Catalog No. NC1094586 Supplier Zymo Research Corporation Supplier No. C100750
It is ideal for clarifying solutions, including crude cell lysates and homogenates. The silica filtration membrane has an approximate 10 - 20 m pore size. Capacity is 700 l.