Learn More
Zymo Research Corporation Zymo-Spin™ IV Columns (250 Pack)
SDP
Catalog No. 501251279
Encompass_Preferred
It is ideal for clarifying solutions, including crude cell lysates and homogenates. The silica filtration membrane has an approximate 10 - 20 m pore size. Capacity is 700 l.
Catalog No. 50-125-1279 Supplier Zymo Research Corporation Supplier No. C1007250
It is ideal for clarifying solutions, including crude cell lysates and homogenates. The silica filtration membrane has an approximate 10 - 20 m pore size. Capacity is 700 l.