Learn More
Zymo Research Corporation Zymo-Spin™ V Columns (50 Pack) (Uncapped)
SDP
Catalog No. 501251283
Encompass_Preferred
This column features a luer-lock top allowing it to be easily attached to a syringe. The Zymo-Spin V features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 100 g DNA or RNA in 150 l eluate. Capacity is 800 l.
Catalog No. 50-125-1283 Supplier Zymo Research Corporation Supplier No. C101250
This column features a luer-lock top allowing it to be easily attached to a syringe. The Zymo-Spin V features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 100 g DNA or RNA in 150 l eluate. Capacity is 800 l.