Learn More
Zymo Research Corporation Zymo-Spin™ V-E Columns (50 Pack)
SDP
Catalog No. 501251293
Encompass_Preferred
This column features a luer-lock top allowing it to be easily attached to a syringe, resesrvoir, or prefilter. The Zymo-Spin V-E features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 125 g DNA, and 250 g RNA, in 150 l eluate. Capacity is 400 l.
Catalog No. 50-125-1293 Supplier Zymo Research Corporation Supplier No. C102450
This column features a luer-lock top allowing it to be easily attached to a syringe, resesrvoir, or prefilter. The Zymo-Spin V-E features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 125 g DNA, and 250 g RNA, in 150 l eluate. Capacity is 400 l.