Learn More
Zymo Research Corporation Zymo-Spin V-PS Column Assembly w/15 ml Reservoir-X and 50 ml Reservoir
SDP
Catalog No. NC2144350
Encompass_Preferred
Available as a refill for the ZymoPURE II Plasmid Midiprep and ZymoPURE – Express Plasmid Midiprep Kits. The versatile Zymo-Spin V-PS with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PS features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 1.2 mg of plasmid DNA in ≥ 150 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.
Catalog No. NC2144350 Supplier Zymo Research Corporation Supplier No. C10835
Available as a refill for the ZymoPURE II Plasmid Midiprep and ZymoPURE – Express Plasmid Midiprep Kits. The versatile Zymo-Spin V-PS with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PS features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 1.2 mg of plasmid DNA in ≥ 150 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.