Learn More
Zymo Research Corporation Zymo-Spin V-PX Column Assembly w/15 ml Reservoir-X and 50 ml Reservoir
SDP
Catalog No. NC2144349
Encompass_Preferred
Available as a refill for the ZymoPURE II Plasmid Maxiprep Kit. The versatile Zymo-Spin V-PX with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 3 mg of plasmid DNA in ≥ 300 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.
This item is not returnable. View return policy
Catalog No. NC2144349 Supplier Zymo Research Corporation Supplier No. C10825
Available as a refill for the ZymoPURE II Plasmid Maxiprep Kit. The versatile Zymo-Spin V-PX with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 3 mg of plasmid DNA in ≥ 300 µl eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.