Learn More
Zymo Research Corporation Zymo-Spin™ VI Columns (20 Pack) (Uncapped)
SDP
Catalog No. 502036993
Encompass_Preferred
Exclusive to this column is a luer-lock bottom assembly. The Zymo-Spin VI features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 500 g DNA or RNA in 2 ml eluate. Capacity is 15 ml.
Catalog No. 50-203-6993 Supplier Zymo Research Corporation Supplier No. C101320
Exclusive to this column is a luer-lock bottom assembly. The Zymo-Spin VI features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 500 g DNA or RNA in 2 ml eluate. Capacity is 15 ml.