Learn More
Zymo Research Corporation Zymo-Spin™ VI Columns w/ Zymo Maxi Filter™ (20 Pack)
SDP
Catalog No. 502036999
Encompass_Preferred
The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 g DNA or RNA in 2 ml eluate. The capacity of the spin-column with filter is 75 ml.
Catalog No. 50-203-6999 Supplier Zymo Research Corporation Supplier No. C101720
The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 g DNA or RNA in 2 ml eluate. The capacity of the spin-column with filter is 75 ml.