Learn More
Research Products International Corp Zymo-Spin VI-PX Column 5pk
SDP
Catalog No. 502388460
Encompass
Available as a refill for the ZymoPURE II Plasmid Gigaprep Kit. The versatile Zymo-Spin VI-PX can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. Exclusive to this column is a Luer-Lock bottom assembly and conical tip. The Zymo-Spin VI-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 25 mg of plasmid DNA in ≥ 3 ml eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 15 ml and can be increased to 615 ml when used in conjunction with the 600 ml reservoirZC1033-5.
Catalog No. 50-238-8460 Supplier Research Products International Corp Supplier No. ZC10805
Available as a refill for the ZymoPURE II Plasmid Gigaprep Kit. The versatile Zymo-Spin VI-PX can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. Exclusive to this column is a Luer-Lock bottom assembly and conical tip. The Zymo-Spin VI-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 25 mg of plasmid DNA in ≥ 3 ml eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 15 ml and can be increased to 615 ml when used in conjunction with the 600 ml reservoirZC1033-5.