Learn More
Zymo Research Corporation Zymo-Spin VI-PX (5 pack)
SDP
Catalog No. NC2144348
Encompass_Preferred
Available as a refill for the ZymoPURE II Plasmid Gigaprep Kit. The versatile Zymo-Spin VI-PX can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. Exclusive to this column is a Luer-Lock bottom assembly and conical tip. The Zymo-Spin VI-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 25 mg of plasmid DNA in 3 ml eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 15 ml and can be increased to 615 ml when used in conjunction with the 600 ml reservoir (Cat No. C1033-5).
This item is not returnable. View return policy
Catalog No. NC2144348 Supplier Zymo Research Corporation Supplier No. C10805
Available as a refill for the ZymoPURE II Plasmid Gigaprep Kit. The versatile Zymo-Spin VI-PX can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. Exclusive to this column is a Luer-Lock bottom assembly and conical tip. The Zymo-Spin VI-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 25 mg of plasmid DNA in 3 ml eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity is 15 ml and can be increased to 615 ml when used in conjunction with the 600 ml reservoir (Cat No. C1033-5).