Learn More
Zymo Research Corporation Zymo-Spin™ II Columns (50 Pack) (Uncapped)
SDP
Catalog No. 50444051
Encompass_Preferred
The Zymo-Spin™ II unique column features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g DNA or RNA in 25 l eluate. Capacity is 800 l.
Catalog No. 50-444-051 Supplier Zymo Research Corporation Supplier No. C100850
The Zymo-Spin™ II unique column features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g DNA or RNA in 25 l eluate. Capacity is 800 l.