Learn More
Zymo Research Corporation YD Digestion Buffer, 4.8ml
SDP
Catalog No. NC0351055
Encompass_Preferred
YD Digestion Buffer provides a reliable and efficient method for the isolation of Genomic DNA from a broad spectrum of fungus species using Zymolyase and Zymo-Spin column purification technology.
Catalog No. NC0351055 Supplier Zymo Research Corporation Supplier No. D20021
YD Digestion Buffer provides a reliable and efficient method for the isolation of Genomic DNA from a broad spectrum of fungus species using Zymolyase and Zymo-Spin column purification technology.